ENA428074144 Enamine

4-(2-methylpropyl)benzene-1,3-diol

  • MDL number MFCD24713582
  • Cas Number : 18979-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180811
Availability details for
Quantity
  • Availability
  • Add to Cart