ENA428074157 Enamine

N-cyclopentyl-2-methylpropanamide

  • MDL number MFCD01008125
  • Cas Number : 349130-19-4
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-178753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178753
Availability details for
Quantity
  • Availability
  • Add to Cart