ENA428074165 Enamine

1-(4-nitro-3-propoxyphenyl)piperidin-4-ol

  • MDL number MFCD28064027
  • Cas Number : 1409764-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180858
Availability details for
Quantity
  • Availability
  • Add to Cart