ENA428074169 Enamine

(1S)-1-[3-(benzyloxy)phenyl]ethan-1-ol

  • MDL number MFCD18340263
  • Cas Number : 1280591-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180869
Availability details for
Quantity
  • Availability
  • Add to Cart