ENA428074188 Enamine

1-N,2-N,4-tribenzylazetidine-2,4-dicarboxamide

  • MDL number MFCD28064037
  • Cas Number : 1786096-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-180953
Availability details for
Quantity
  • Availability
  • Add to Cart