ENA428074189 Enamine

2-(aminomethyl)-6-bromophenol hydrochloride

  • MDL number MFCD10566809
  • Cas Number : 1795488-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180954
Availability details for
Quantity
  • Availability
  • Add to Cart