ENA428074192 Enamine

3-aminopyrazine-2-thiol

  • MDL number MFCD18449265
  • Cas Number : 31613-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-250
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180973
Availability details for
Quantity
  • Availability
  • Add to Cart