ENA428074193 Enamine

methyl(2-methylbut-3-yn-2-yl)amine

  • MDL number MFCD19630049
  • Cas Number : 2978-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180974
Availability details for
Quantity
  • Availability
  • Add to Cart