ENA428074197 Enamine

1-[1-(morpholin-4-yl)cyclopropyl]methanamine

  • MDL number MFCD26937946
  • Cas Number : 1695222-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180982
Availability details for
Quantity
  • Availability
  • Add to Cart