ENA428074201 Enamine

2-[(cyclopropylmethyl)carbamoyl]acetic acid

  • MDL number MFCD06208737
  • Cas Number : 773099-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178829
Availability details for
Quantity
  • Availability
  • Add to Cart