ENA428074206 Enamine

3-methyl-N-(propan-2-yl)-1,2,4-oxadiazol-5-amine

  • MDL number MFCD16818096
  • Cas Number : 1251075-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181043
Availability details for
Quantity
  • Availability
  • Add to Cart