ENA428074210 Enamine

4-[2-(trifluoromethyl)phenyl]piperidin-4-ol hydrochloride

  • MDL number MFCD28064040
  • Cas Number : 208989-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181057
Availability details for
Quantity
  • Availability
  • Add to Cart