ENA428074212 Enamine

4-(2-chlorophenyl)piperidin-4-ol hydrochloride

  • MDL number MFCD28064041
  • Cas Number : 860228-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181068
Availability details for
Quantity
  • Availability
  • Add to Cart