ENA428074218 Enamine

(2S)-2-(morpholin-4-yl)propan-1-ol

  • MDL number MFCD26906050
  • Cas Number : 1352306-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181084
Availability details for
Quantity
  • Availability
  • Add to Cart