ENA428074219 Enamine

2-(2-ethoxyethyl)oxirane

  • MDL number MFCD22414164
  • Cas Number : 99115-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181086
Availability details for
Quantity
  • Availability
  • Add to Cart