ENA428074223 Enamine

methyl 2-chloro-5-methyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD16660888
  • Cas Number : 1378819-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181104
Availability details for
Quantity
  • Availability
  • Add to Cart