ENA428074228 Enamine

6-(hydroxymethyl)-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD19646925
  • Cas Number : 207552-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181114
Availability details for
Quantity
  • Availability
  • Add to Cart