ENA428074229 Enamine

ethyl 1,2,3-thiadiazole-5-carboxylate

  • MDL number MFCD00971933
  • Cas Number : 4100-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181116
Availability details for
Quantity
  • Availability
  • Add to Cart