ENA428074236 Enamine

methyl 4-bromo-2-methoxy-5-sulfamoylbenzoate

  • MDL number MFCD24189674
  • Cas Number : 1236476-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181130
Availability details for
Quantity
  • Availability
  • Add to Cart