ENA428074239 Enamine

1-(2-methylpyrimidin-4-yl)ethan-1-one

  • MDL number MFCD00209495
  • Cas Number : 67860-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178845
Availability details for
Quantity
  • Availability
  • Add to Cart