ENA428074244 Enamine

N3-ethyl-4-[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]-1H-pyrazole-3,5-diamine

  • MDL number MFCD14729561
  • Cas Number : 1188305-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181266
Availability details for
Quantity
  • Availability
  • Add to Cart