ENA428074245 Enamine

4-[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]-N3-methyl-1H-pyrazole-3,5-diamine

  • MDL number MFCD14729562
  • Cas Number : 1188305-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181267
Availability details for
Quantity
  • Availability
  • Add to Cart