ENA428074247 Enamine

7-benzyl-2-(4-fluorophenyl)-3H,4H,5H,6H,7H,8H-pyrido[3,4-d]pyrimidin-4-one

  • MDL number MFCD14729816
  • Cas Number : 1031970-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181306
Availability details for
Quantity
  • Availability
  • Add to Cart