ENA428074249 Enamine

phenyl N-(3-chlorophenyl)carbamate

  • MDL number MFCD00018424
  • Cas Number : 50699-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178847
Availability details for
Quantity
  • Availability
  • Add to Cart