ENA428074250 Enamine

5-ethyl-1-(3-methylbutyl)-1H-pyrazol-4-amine

  • MDL number MFCD21773967
  • Cas Number : 1536848-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178848
Availability details for
Quantity
  • Availability
  • Add to Cart