ENA428074256 Enamine

2-amino-N-[(1,3-dioxaindan-5-yl)methyl]-6-chlorobenzamide

  • MDL number MFCD28063905
  • Cas Number : 1411540-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178865
Availability details for
Quantity
  • Availability
  • Add to Cart