ENA428074257 Enamine

4-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]-3-(methylsulfanyl)-1H-pyrazol-5-amine

  • MDL number MFCD26962828
  • Cas Number : 1188304-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181410
Availability details for
Quantity
  • Availability
  • Add to Cart