ENA428074260 Enamine

6-chloro-2,3-dimethylaniline hydrochloride

  • MDL number MFCD28064058
  • Cas Number : 1798732-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181423
Availability details for
Quantity
  • Availability
  • Add to Cart