ENA428074263 Enamine

1-(4-cyclobutylphenyl)methanamine

  • MDL number MFCD28063906
  • Cas Number : 1553955-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178868
Availability details for
Quantity
  • Availability
  • Add to Cart