ENA428074264 Enamine

4-amino-N-(5-bromo-2-hydroxyphenyl)-3-methylbenzamide

  • MDL number MFCD24790696
  • Cas Number : 1522495-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178474
Availability details for
Quantity
  • Availability
  • Add to Cart