ENA428074265 Enamine

2-ethylbutanamide

  • MDL number MFCD00025512
  • Cas Number : 1114-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181430
Availability details for
Quantity
  • Availability
  • Add to Cart