ENA428074275 Enamine

4-propyl-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD17251109
  • Cas Number : 211942-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178873
Availability details for
Quantity
  • Availability
  • Add to Cart