ENA428074277 Enamine

1-bromocycloocta-1,5-diene

  • Cas Number : 57559-44-1
  • Molecular Weight : 187.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181490
Availability details for
Quantity
  • Availability
  • Add to Cart