ENA428074278 Enamine

5-(chloromethyl)-1-[(oxan-4-yl)methyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD18869500
  • Cas Number : 1340063-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181493
Availability details for
Quantity
  • Availability
  • Add to Cart