ENA428074279 Enamine

[(4-bromo-2-methylphenyl)methyl](methyl)amine

  • MDL number MFCD12096719
  • Cas Number : 1037090-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181494
Availability details for
Quantity
  • Availability
  • Add to Cart