ENA428074281 Enamine

[(5-chloro-2-fluorophenyl)methyl](methyl)amine

  • MDL number MFCD11105973
  • Cas Number : 1152846-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181496
Availability details for
Quantity
  • Availability
  • Add to Cart