ENA428074293 Enamine

2-amino-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD03840776
  • Cas Number : 76391-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181552
Availability details for
Quantity
  • Availability
  • Add to Cart