ENA428074296 Enamine

1-(2-ethyl-3-hydroxy-2-methylpropyl)pyrrolidin-3-ol

  • MDL number MFCD17249618
  • Cas Number : 1344288-73-8
  • Molecular Weight : 187.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181562
Availability details for
Quantity
  • Availability
  • Add to Cart