ENA428074297 Enamine

tert-butyl 4-hydroxy-5H,6H,7H,8H-pyrido[3,4-d]pyrimidine-7-carboxylate

  • MDL number MFCD11869803
  • Cas Number : 1142188-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178477
Availability details for
Quantity
  • Availability
  • Add to Cart