ENA428074298 Enamine

but-3-yn-2-yl methanesulfonate

  • MDL number MFCD20039790
  • Cas Number : 59967-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181568
Availability details for
Quantity
  • Availability
  • Add to Cart