ENA428074300 Enamine

(1R)-1-[3-(benzyloxy)phenyl]ethan-1-ol

  • MDL number MFCD18340336
  • Cas Number : 1354485-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181578
Availability details for
Quantity
  • Availability
  • Add to Cart