ENA428074310 Enamine

2-(2,2,2-trifluoroethoxy)propanenitrile

  • MDL number MFCD16079400
  • Cas Number : 1250815-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181600
Availability details for
Quantity
  • Availability
  • Add to Cart