ENA428074313 Enamine

5-ethyl-1-[(4-fluorophenyl)methyl]-1H-1,2,3-triazol-4-amine hydrochloride

  • MDL number MFCD28063995
  • Cas Number : 1798728-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181618
Availability details for
Quantity
  • Availability
  • Add to Cart