ENA428074317 Enamine

(1S)-1-[3-(benzyloxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD12756871
  • Cas Number : 1391541-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181622
Availability details for
Quantity
  • Availability
  • Add to Cart