ENA428074320 Enamine

2-chlorofuran-3-carboxylic acid

  • MDL number MFCD18393139
  • Cas Number : 31491-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181629
Availability details for
Quantity
  • Availability
  • Add to Cart