ENA428074332 Enamine

6-hydrazinyl-2,4-dimethyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD12103337
  • Cas Number : 54317-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181694
Availability details for
Quantity
  • Availability
  • Add to Cart