ENA428074333 Enamine

[2-(trifluoromethyl)-1,3-thiazol-5-yl]methanol

  • MDL number MFCD18632535
  • Cas Number : 131748-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181697
Availability details for
Quantity
  • Availability
  • Add to Cart