ENA428074341 Enamine

6-fluoro-3-methyl-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD02741583
  • Cas Number : 130507-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181705
Availability details for
Quantity
  • Availability
  • Add to Cart