ENA428074342 Enamine

methyl 3-bromo-5-chloro-2-fluorobenzoate

  • MDL number MFCD24107111
  • Cas Number : 1353575-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181724
Availability details for
Quantity
  • Availability
  • Add to Cart