ENA428074346 Enamine

2,3-dihydro-1H-indole-1-carboximidamide

  • MDL number MFCD06612529
  • Cas Number : 59589-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178910
Availability details for
Quantity
  • Availability
  • Add to Cart